The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015340.3 | NP_056155.1 | 11 - 12 | 1308 | 65 |
XM_005265006.1 | XP_005265063.1 | 10 - 11 | 1264 | 65 | |
XM_011533554.2 | XP_011531856.1 | 11 - 12 | 1327 | 65 | |
XM_017006042.1 | XP_016861531.1 | 11 - 12 | 1327 | 65 | |
GenBank mRNA | AK025465.1 | - | 5 - 6 | 608 | 65 |
AK308146.1 | - | 9 - 10 | 1132 | 65 | |
BC025989.2 | - | 11 - 12 | 1315 | 65 | |
D21851.1 | - | 11 - 12 | 1308 | 65 |