The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015340.3 | NP_056155.1 | 16 - 17 | 2043 | 95 |
XM_005265006.1 | XP_005265063.1 | 15 - 16 | 1999 | 95 | |
XM_011533554.2 | XP_011531856.1 | 16 - 17 | 2062 | 95 | |
XM_017006042.1 | XP_016861531.1 | 16 - 17 | 2062 | 95 | |
GenBank mRNA | AK025465.1 | - | 10 - 11 | 1343 | 95 |
AK026113.1 | - | 3 - 4 | 266 | 95 | |
AK308146.1 | - | 14 - 15 | 1867 | 95 | |
BC025989.2 | - | 16 - 17 | 2050 | 95 | |
D21851.1 | - | 16 - 17 | 2043 | 95 |