The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015340.3 | NP_056155.1 | 17 - 18 | 2231 | 82 |
XM_005265006.1 | XP_005265063.1 | 16 - 17 | 2187 | 82 | |
XM_011533554.2 | XP_011531856.1 | 17 - 18 | 2250 | 82 | |
XM_017006042.1 | XP_016861531.1 | 17 - 18 | 2250 | 82 | |
GenBank mRNA | AK025465.1 | - | 11 - 12 | 1531 | 82 |
AK026113.1 | - | 4 - 5 | 454 | 82 | |
AK308146.1 | - | 15 - 16 | 2055 | 82 | |
BC025989.2 | - | 17 - 18 | 2238 | 82 | |
D21851.1 | - | 17 - 18 | 2231 | 82 |