The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 11 - 12 | 1552 | 137 |
XM_011511183.2 | XP_011509485.1 | 11 - 12 | 1553 | 137 | |
GenBank mRNA | AK295005.1 | - | 11 - 12 | 1474 | 137 |
AY265357.1 | - | 11 - 12 | 1406 | 137 | |
AY265358.1 | - | 11 - 12 | 1403 | 137 | |
U33837.1 | - | 11 - 12 | 1443 | 137 |