The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 45 - 46 | 8794 | 105 |
XM_011511183.2 | XP_011509485.1 | 45 - 46 | 8795 | 105 | |
XM_011511184.2 | XP_011509486.1 | 30 - 31 | 6307 | 105 | |
GenBank mRNA | AY265357.1 | - | 45 - 46 | 8648 | 105 |
AY265358.1 | - | 45 - 46 | 8645 | 105 |