The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 55 - 56 | 10978 | 67 |
XM_011511183.2 | XP_011509485.1 | 55 - 56 | 10979 | 67 | |
XM_011511184.2 | XP_011509486.1 | 40 - 41 | 8491 | 67 | |
GenBank mRNA | AK302843.1 | - | 6 - 7 | 968 | 67 |
AY265357.1 | - | 55 - 56 | 10832 | 67 | |
AY265358.1 | - | 55 - 56 | 10829 | 67 | |
U04441.1 | - | 13 - 14 | 2656 | 67 | |
U33837.1 | - | 55 - 56 | 10869 | 67 |