The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032221.3 | NP_001027392.1 | 10 - 11 | 1095 | 100 |
NM_003165.3 | NP_003156.1 | 10 - 11 | 1095 | 100 | |
GenBank mRNA | AB209180.1 | - | 4 - 5 | 359 | 100 |
AF004562.1 | - | 10 - 11 | 1018 | 100 | |
AF004563.1 | - | 10 - 11 | 1018 | 100 | |
AK293984.1 | - | 9 - 10 | 914 | 100 | |
AK295154.1 | - | 9 - 10 | 953 | 100 | |
AK295802.1 | - | 5 - 6 | 521 | 100 | |
BC015749.1 | - | 10 - 11 | 1011 | 100 | |
D63851.1 | - | 10 - 11 | 967 | 100 | |
Y12723.1 | - | 10 - 11 | 1006 | 100 |