The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002843.3 | NP_002834.3 | 19 - 20 | 3588 | 86 |
XM_017018083.1 | XP_016873572.1 | 18 - 19 | 3319 | 86 | |
XM_017018084.1 | XP_016873573.1 | 19 - 20 | 3701 | 86 | |
XM_017018085.1 | XP_016873574.1 | 19 - 20 | 3200 | 86 | |
GenBank mRNA | D37781.1 | - | 19 - 20 | 3577 | 86 |
U10886.1 | - | 19 - 20 | 3582 | 86 |