The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198182.2 | NP_937825.2 | 12 - 13 | 1668 | 90 |
XM_005246159.3 | XP_005246216.1 | 11 - 12 | 1554 | 90 | |
XM_006711882.2 | XP_006711945.1 | 12 - 13 | 1704 | 90 | |
XM_006711884.3 | XP_006711947.1 | 11 - 12 | 1583 | 90 | |
XM_011510343.2 | XP_011508645.1 | 12 - 13 | 1675 | 90 | |
XM_017003900.1 | XP_016859389.1 | 12 - 13 | 1777 | 90 | |
XM_017003901.1 | XP_016859390.1 | 12 - 13 | 1741 | 90 | |
GenBank mRNA | AF198489.1 | - | 12 - 13 | 1606 | 90 |
AF411210.1 | - | 8 - 9 | 1023 | 90 | |
AK293855.1 | - | 12 - 13 | 1715 | 90 | |
AK296982.1 | - | 12 - 13 | 1746 | 90 | |
AK312950.1 | - | 12 - 13 | 1638 | 90 | |
BC067519.1 | - | 12 - 13 | 1549 | 90 | |
BC067520.1 | - | 12 - 13 | 1549 | 90 | |
BC067521.1 | - | 12 - 13 | 1575 | 90 |