The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141973.2 | NP_001135445.1 | 13 - 14 | 1524 | 101 |
NM_001141974.2 | NP_001135446.1 | 13 - 14 | 1524 | 101 | |
NM_022089.3 | NP_071372.1 | 13 - 14 | 1539 | 101 | |
XM_005245810.1 | XP_005245867.1 | 13 - 14 | 1536 | 101 | |
XM_005245811.1 | XP_005245868.1 | 13 - 14 | 1524 | 101 | |
XM_005245812.1 | XP_005245869.1 | 13 - 14 | 1512 | 101 | |
XM_005245815.1 | XP_005245872.1 | 13 - 14 | 1539 | 101 | |
XM_006710512.1 | XP_006710575.1 | 13 - 14 | 1521 | 101 | |
XM_006710513.1 | XP_006710576.1 | 13 - 14 | 1497 | 101 | |
XM_011541128.1 | XP_011539430.1 | 13 - 14 | 1539 | 101 | |
XM_011541129.1 | XP_011539431.1 | 13 - 14 | 1539 | 101 | |
XM_017000844.1 | XP_016856333.1 | 13 - 14 | 1539 | 101 | |
XM_017000845.1 | XP_016856334.1 | 13 - 14 | 1521 | 101 | |
XM_017000846.1 | XP_016856335.1 | 13 - 14 | 1497 | 101 | |
XM_017000847.1 | XP_016856336.1 | 13 - 14 | 1509 | 101 | |
XM_017000848.1 | XP_016856337.1 | 13 - 14 | 1539 | 101 | |
XM_017000849.1 | XP_016856338.1 | 13 - 14 | 1524 | 101 | |
XM_017000850.1 | XP_016856339.1 | 13 - 14 | 1539 | 101 | |
GenBank mRNA | AK075310.1 | - | 3 - 4 | 321 | 101 |
AK290210.1 | - | 13 - 14 | 1480 | 101 | |
AL354615.1 | - | 13 - 14 | 1339 | 101 | |
AY461712.1 | - | 13 - 14 | 1338 | 101 | |
BC030267.1 | - | 13 - 14 | 1467 | 101 | |
BC034575.1 | - | 10 - 11 | 1030 | 101 |