Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 23 - 24 2826 74
NM_001141974.2 NP_001135446.1 22 - 23 2709 74
NM_022089.3 NP_071372.1 23 - 24 2841 74
XM_005245810.1 XP_005245867.1 23 - 24 2838 74
XM_005245811.1 XP_005245868.1 23 - 24 2826 74
XM_005245812.1 XP_005245869.1 23 - 24 2814 74
XM_005245815.1 XP_005245872.1 22 - 23 2724 74
XM_006710512.1 XP_006710575.1 23 - 24 2823 74
XM_006710513.1 XP_006710576.1 23 - 24 2799 74
XM_011541128.1 XP_011539430.1 23 - 24 2826 74
XM_011541129.1 XP_011539431.1 22 - 23 2634 74
XM_017000844.1 XP_016856333.1 23 - 24 2826 74
XM_017000845.1 XP_016856334.1 23 - 24 2823 74
XM_017000846.1 XP_016856335.1 23 - 24 2799 74
XM_017000847.1 XP_016856336.1 23 - 24 2796 74
XM_017000848.1 XP_016856337.1 22 - 23 2724 74
XM_017000849.1 XP_016856338.1 22 - 23 2709 74
XM_017000850.1 XP_016856339.1 22 - 23 2634 74
GenBank mRNA AJ009947.1 - 1 - 2 47 74
AK025928.1 - 3 - 4 230 74
AK290210.1 - 23 - 24 2782 74
AL354615.1 - 23 - 24 2641 74
AL833966.1 - 4 - 5 495 74
AY461712.1 - 23 - 24 2640 74
BC030267.1 - 22 - 23 2652 74
BC034575.1 - 20 - 21 2332 74

View Full Product Details