Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 6 - 7 775 78
NM_001141974.2 NP_001135446.1 6 - 7 775 78
NM_022089.3 NP_071372.1 6 - 7 790 78
XM_005245810.1 XP_005245867.1 6 - 7 787 78
XM_005245811.1 XP_005245868.1 6 - 7 775 78
XM_005245812.1 XP_005245869.1 6 - 7 790 78
XM_005245815.1 XP_005245872.1 6 - 7 790 78
XM_006710512.1 XP_006710575.1 6 - 7 772 78
XM_006710513.1 XP_006710576.1 6 - 7 775 78
XM_011541128.1 XP_011539430.1 6 - 7 790 78
XM_011541129.1 XP_011539431.1 6 - 7 790 78
XM_017000844.1 XP_016856333.1 6 - 7 790 78
XM_017000845.1 XP_016856334.1 6 - 7 772 78
XM_017000846.1 XP_016856335.1 6 - 7 775 78
XM_017000847.1 XP_016856336.1 6 - 7 787 78
XM_017000848.1 XP_016856337.1 6 - 7 790 78
XM_017000849.1 XP_016856338.1 6 - 7 775 78
XM_017000850.1 XP_016856339.1 6 - 7 790 78
GenBank mRNA AK290210.1 - 6 - 7 731 78
AL354615.1 - 6 - 7 590 78
AY461712.1 - 6 - 7 589 78
BC030267.1 - 6 - 7 718 78
BC034575.1 - 3 - 4 281 78

View Full Product Details