Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 19 - 20 2105 116
NM_001313727.1 NP_001300656.1 15 - 16 1663 116
NM_031418.3 NP_113606.2 18 - 19 2013 116
XM_011520282.2 XP_011518584.1 5 - 6 560 116
XM_017018118.1 XP_016873607.1 16 - 17 1641 116
XM_017018119.1 XP_016873608.1 13 - 14 1253 116
GenBank mRNA AJ300461.1 - 18 - 19 2720 116
AK295816.1 - 15 - 16 1575 116
AK302659.1 - 18 - 19 2052 116
AK304781.1 - 19 - 20 1946 116
AK316018.1 - 15 - 16 1575 116

View Full Product Details