The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015316.2 | NP_056131.2 | 14 - 15 | 3150 | 72 |
XM_005267487.4 | XP_005267544.1 | 14 - 15 | 3554 | 72 | |
XM_011536593.2 | XP_011534895.1 | 15 - 16 | 3659 | 72 | |
XM_017021116.1 | XP_016876605.1 | 15 - 16 | 3296 | 72 | |
XM_017021117.1 | XP_016876606.1 | 15 - 16 | 3072 | 72 | |
XM_017021118.1 | XP_016876607.1 | 15 - 16 | 2985 | 72 | |
XM_017021119.1 | XP_016876608.1 | 9 - 10 | 2279 | 72 | |
GenBank mRNA | AB018314.1 | - | 10 - 11 | 2441 | 72 |
AJ318887.2 | - | 14 - 15 | 3025 | 72 | |
AK125913.1 | - | 16 - 17 | 3521 | 72 | |
AK307149.1 | - | 2 - 3 | 524 | 72 | |
BC035446.1 | - | 14 - 15 | 3119 | 72 | |
BC136527.1 | - | 14 - 15 | 2954 | 72 |