Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
Chromosome Location
Chr.5: 41925249 - 42040230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297437.1 NP_001284366.1 4 - 5 771 121
NM_012176.2 NP_036308.1 4 - 5 769 121
NM_033484.2 NP_277019.1 4 - 5 769 121
XM_006714470.3 XP_006714533.1 3 - 4 467 121
XM_011514026.2 XP_011512328.1 4 - 5 774 121
XM_011514027.2 XP_011512329.1 4 - 5 2144 121
GenBank mRNA AF129534.1 - 4 - 5 713 121
AF176703.1 - 4 - 5 707 121
AK024075.1 - 4 - 5 769 121
BC040011.1 - 4 - 5 730 121
BC048098.1 - 4 - 5 737 121
CR749719.1 - 2 - 3 286 121

View Full Product Details