Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCA
Entrez Gene ID
5095
Gene Name
propionyl-CoA carboxylase alpha subunit
Gene Aliases
-
Chromosome Location
Chr.13: 100089015 - 100530437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000282.3 NP_000273.2 11 - 12 1019 92
NM_001127692.2 NP_001121164.1 10 - 11 941 92
NM_001178004.1 NP_001171475.1 11 - 12 1019 92
XM_005254059.2 XP_005254116.1 11 - 12 1014 92
XM_011521093.1 XP_011519395.1 11 - 12 1014 92
XM_017020605.1 XP_016876094.1 11 - 12 1014 92
XM_017020606.1 XP_016876095.1 10 - 11 865 92
XM_017020607.1 XP_016876096.1 11 - 12 830 92
XM_017020608.1 XP_016876097.1 11 - 12 1014 92
XM_017020609.1 XP_016876098.1 11 - 12 927 92
XM_017020610.1 XP_016876099.1 9 - 10 1281 92
XM_017020611.1 XP_016876100.1 11 - 12 1014 92
XM_017020612.1 XP_016876101.1 11 - 12 1014 92
XM_017020613.1 XP_016876102.1 11 - 12 1014 92
XM_017020614.1 XP_016876103.1 10 - 11 865 92
XM_017020615.1 XP_016876104.1 11 - 12 1014 92
XM_017020616.1 XP_016876105.1 11 - 12 1014 92
XM_017020617.1 XP_016876106.1 7 - 8 598 92
XM_017020618.1 XP_016876107.1 7 - 8 598 92
GenBank mRNA AF385926.1 - 11 - 12 951 92
AK225299.1 - 11 - 12 917 92
AK296771.1 - 10 - 11 839 92
AK298318.1 - 11 - 12 941 92
AK315503.1 - 11 - 12 917 92
BC000140.2 - 11 - 12 891 92
X14608.1 - 11 - 12 886 92

View Full Product Details