Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCA
Entrez Gene ID
5095
Gene Name
propionyl-CoA carboxylase alpha subunit
Gene Aliases
-
Chromosome Location
Chr.13: 100089015 - 100530437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000282.3 NP_000273.2 4 - 5 408 65
NM_001127692.2 NP_001121164.1 3 - 4 330 65
NM_001178004.1 NP_001171475.1 4 - 5 408 65
XM_005254059.2 XP_005254116.1 4 - 5 403 65
XM_011521093.1 XP_011519395.1 4 - 5 403 65
XM_017020605.1 XP_016876094.1 4 - 5 403 65
XM_017020606.1 XP_016876095.1 3 - 4 254 65
XM_017020607.1 XP_016876096.1 4 - 5 219 65
XM_017020608.1 XP_016876097.1 4 - 5 403 65
XM_017020609.1 XP_016876098.1 4 - 5 316 65
XM_017020611.1 XP_016876100.1 4 - 5 403 65
XM_017020612.1 XP_016876101.1 4 - 5 403 65
XM_017020613.1 XP_016876102.1 4 - 5 403 65
XM_017020614.1 XP_016876103.1 3 - 4 254 65
XM_017020615.1 XP_016876104.1 4 - 5 403 65
XM_017020616.1 XP_016876105.1 4 - 5 403 65
XM_017020617.1 XP_016876106.1 1 - 2 66 65
XM_017020618.1 XP_016876107.1 1 - 2 66 65
GenBank mRNA AF385926.1 - 4 - 5 340 65
AK225288.1 - 4 - 5 308 65
AK225299.1 - 4 - 5 306 65
AK298318.1 - 4 - 5 330 65
AK315503.1 - 4 - 5 306 65
BC000140.2 - 4 - 5 280 65
DA781564.1 - 3 - 4 228 65
X14608.1 - 4 - 5 275 65

View Full Product Details