The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206540.2 | NP_001193469.1 | 4 - 5 | 530 | 64 |
NM_001206541.2 | NP_001193470.1 | 5 - 6 | 1027 | 64 | |
NM_001282162.1 | NP_001269091.1 | 4 - 5 | 842 | 64 | |
NM_004930.4 | NP_004921.1 | 4 - 5 | 530 | 64 | |
XM_006710938.3 | XP_006711001.1 | 4 - 5 | 423 | 64 | |
XM_011542228.2 | XP_011540530.1 | 4 - 5 | 1351 | 64 | |
XM_011542229.2 | XP_011540531.1 | 3 - 4 | 538 | 64 | |
XM_011542230.2 | XP_011540532.1 | 3 - 4 | 513 | 64 | |
XM_017002428.1 | XP_016857917.1 | 4 - 5 | 395 | 64 | |
GenBank mRNA | AK125085.1 | - | 5 - 6 | 899 | 64 |
AK301442.1 | - | 4 - 5 | 844 | 64 | |
AK313113.1 | - | 4 - 5 | 366 | 64 | |
BC024601.1 | - | 1 - 2 | 79 | 64 | |
BC107752.1 | - | 4 - 5 | 376 | 64 | |
BC109241.1 | - | 3 - 4 | 327 | 64 | |
BC109242.1 | - | 3 - 4 | 327 | 64 | |
BE870234.1 | - | 4 - 5 | 387 | 64 | |
BT019470.1 | - | 3 - 4 | 327 | 64 | |
BT019471.1 | - | 3 - 4 | 327 | 64 | |
CB267021.1 | - | 1 - 2 | 121 | 64 | |
U03271.1 | - | 3 - 4 | 327 | 64 |