The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076552.2 | NP_001070020.2 | 4 - 5 | 696 | 80 |
NM_001242393.1 | NP_001229322.1 | 4 - 5 | 572 | 80 | |
NM_018677.3 | NP_061147.1 | 4 - 5 | 696 | 80 | |
XM_005260455.1 | XP_005260512.1 | 4 - 5 | 696 | 80 | |
XM_005260456.1 | XP_005260513.1 | 4 - 5 | 696 | 80 | |
XM_006723826.1 | XP_006723889.1 | 4 - 5 | 572 | 80 | |
XM_011528905.1 | XP_011527207.1 | 5 - 6 | 688 | 80 | |
XM_011528906.1 | XP_011527208.1 | 5 - 6 | 688 | 80 | |
XM_011528907.1 | XP_011527209.1 | 5 - 6 | 688 | 80 | |
XM_011528908.1 | XP_011527210.1 | 5 - 6 | 688 | 80 | |
XM_011528909.2 | XP_011527211.1 | 5 - 6 | 574 | 80 | |
XM_011528911.1 | XP_011527213.1 | 4 - 5 | 583 | 80 | |
XM_011528912.2 | XP_011527214.1 | 5 - 6 | 580 | 80 | |
GenBank mRNA | AF263614.1 | - | 4 - 5 | 575 | 80 |
AK000162.1 | - | 4 - 5 | 511 | 80 | |
AK000188.1 | - | 4 - 5 | 649 | 80 | |
AK025990.1 | - | 4 - 5 | 631 | 80 | |
AK092281.1 | - | 4 - 5 | 608 | 80 | |
AL359946.1 | - | 3 - 4 | 406 | 80 | |
BC010141.2 | - | 1 - 2 | 75 | 80 | |
BC012172.1 | - | 4 - 5 | 619 | 80 | |
BC073846.1 | - | 4 - 5 | 460 | 80 | |
BC098422.1 | - | 3 - 4 | 291 | 80 | |
CR749716.1 | - | 3 - 4 | 406 | 80 |