The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007231.2 | NP_001007232.2 | 6 - 7 | 1226 | 57 |
NM_001166276.1 | NP_001159748.1 | 5 - 6 | 1166 | 57 | |
NM_001166277.1 | NP_001159749.1 | 5 - 6 | 1106 | 57 | |
NM_014882.2 | NP_055697.1 | 5 - 6 | 1163 | 57 | |
XM_005264675.2 | XP_005264732.1 | 6 - 7 | 930 | 57 | |
XM_005264676.2 | XP_005264733.1 | 5 - 6 | 814 | 57 | |
XM_011533206.2 | XP_011531508.1 | 6 - 7 | 974 | 57 | |
XM_011533207.2 | XP_011531509.1 | 6 - 7 | 940 | 57 | |
XM_011533208.2 | XP_011531510.1 | 7 - 8 | 991 | 57 | |
XM_011533209.2 | XP_011531511.1 | 3 - 4 | 531 | 57 | |
XM_017005423.1 | XP_016860912.1 | 6 - 7 | 834 | 57 | |
XM_017005424.1 | XP_016860913.1 | 6 - 7 | 831 | 57 | |
XM_017005425.1 | XP_016860914.1 | 5 - 6 | 717 | 57 | |
XM_017005426.1 | XP_016860915.1 | 3 - 4 | 1168 | 57 | |
XM_017005427.1 | XP_016860916.1 | 3 - 4 | 1165 | 57 | |
GenBank mRNA | AK290396.1 | - | 6 - 7 | 904 | 57 |
AK297056.1 | - | 5 - 6 | 984 | 57 | |
AK297157.1 | - | 6 - 7 | 966 | 57 | |
AK298669.1 | - | 5 - 6 | 993 | 57 | |
AK310742.1 | - | 4 - 5 | 726 | 57 | |
AK311669.1 | - | 6 - 7 | 1165 | 57 | |
BC039591.1 | - | 5 - 6 | 1027 | 57 | |
D29642.1 | - | 5 - 6 | 976 | 57 | |
EU794656.1 | - | 5 - 6 | 1163 | 57 |