The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012325.2 | NP_036457.1 | 6 - 7 | 889 | 107 |
XM_011528696.2 | XP_011526998.1 | 6 - 7 | 891 | 107 | |
GenBank mRNA | AK297273.1 | - | 6 - 7 | 738 | 107 |
AK310817.1 | - | 3 - 4 | 294 | 107 | |
BC040410.1 | - | 6 - 7 | 816 | 107 | |
BC106068.1 | - | 6 - 7 | 819 | 107 | |
BC109281.3 | - | 6 - 7 | 813 | 107 | |
U24166.1 | - | 6 - 7 | 814 | 107 |