Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB40A
Entrez Gene ID
142684
Gene Name
RAB40A, member RAS oncogene family
Gene Aliases
RAR2, RAR2A
Chromosome Location
Chr.X: 103493266 - 103519489 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080879.2 NP_543155.2 2 - 3 273 154
GenBank mRNA AF422143.1 - 2 - 3 273 154

View Full Product Details