The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282663.1 | NP_001269592.1 | 12 - 13 | 70 | |
NM_001282664.1 | NP_001269593.1 | 11 - 12 | 70 | ||
NM_001282665.1 | NP_001269594.1 | 11 - 12 | 70 | ||
NM_001282666.1 | NP_001269595.1 | 10 - 11 | 70 | ||
NM_001282667.1 | NP_001269596.1 | 10 - 11 | 70 | ||
NM_014632.3 | NP_055447.1 | 12 - 13 | 70 | ||
XM_011520490.2 | XP_011518792.1 | 10 - 11 | 70 | ||
XM_011520491.2 | XP_011518793.1 | 10 - 11 | 70 | ||
XM_017018589.1 | XP_016874078.1 | 11 - 12 | 70 | ||
XM_017018590.1 | XP_016874079.1 | 11 - 12 | 70 | ||
XM_017018591.1 | XP_016874080.1 | 11 - 12 | 70 | ||
GenBank mRNA | AB018293.1 | - | 12 - 13 | 70 | |
AB110785.1 | - | 10 - 11 | 70 | ||
AB110786.1 | - | 10 - 11 | 70 | ||
AK294845.1 | - | 11 - 12 | 70 | ||
AK302580.1 | - | 3 - 4 | 304 | 70 | |
AK302893.1 | - | 12 - 13 | 1924 | 70 | |
BC044577.1 | - | 12 - 13 | 70 | ||
BX538021.1 | - | 8 - 9 | 70 |