Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF11
Entrez Gene ID
9826
Gene Name
Rho guanine nucleotide exchange factor 11
Gene Aliases
GTRAP48, PDZ-RHOGEF
Chromosome Location
Chr.1: 156934840 - 157046903 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014784.3 NP_055599.1 35 - 36 4650 65
NM_198236.2 NP_937879.1 36 - 37 4770 65
XM_005245629.4 XP_005245686.1 36 - 37 5174 65
XM_005245633.4 XP_005245690.1 26 - 27 3033 65
XM_006711659.2 XP_006711722.1 38 - 39 5222 65
XM_006711660.3 XP_006711723.1 38 - 39 5219 65
XM_006711661.2 XP_006711724.1 37 - 38 5192 65
XM_006711662.2 XP_006711725.1 37 - 38 5189 65
XM_006711663.3 XP_006711726.1 36 - 37 4209 65
XM_006711665.3 XP_006711728.1 37 - 38 5099 65
XM_011510185.2 XP_011508487.1 37 - 38 5207 65
XM_011510186.2 XP_011508488.1 37 - 38 5102 65
XM_011510187.2 XP_011508489.1 37 - 38 3933 65
XM_011510189.2 XP_011508491.1 15 - 16 3238 65
XM_017002920.1 XP_016858409.1 37 - 38 5189 65
XM_017002921.1 XP_016858410.1 35 - 36 5141 65
XM_017002922.1 XP_016858411.1 36 - 37 5069 65
XM_017002923.1 XP_016858412.1 35 - 36 5054 65
XM_017002924.1 XP_016858413.1 35 - 36 5054 65
GenBank mRNA AB002378.1 - 35 - 36 4355 65
AK302500.1 - 15 - 16 65
BC057394.1 - 36 - 37 4794 65

View Full Product Details