Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
Chromosome Location
Chr.6: 32968660 - 32981505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113182.2 NP_001106653.1 10 - 11 3450 86
NM_001199456.1 NP_001186385.1 9 - 10 1853 86
NM_001291986.1 NP_001278915.1 11 - 12 3542 86
NM_005104.3 NP_005095.1 10 - 11 3537 86
GenBank mRNA AL832722.1 - 11 - 12 3477 86
BX647233.1 - 9 - 10 1853 86
BX648109.1 - 10 - 11 3451 86
D42040.1 - 10 - 11 3537 86
M80613.1 - 9 - 10 2872 86
X62083.1 - 9 - 10 2872 86

View Full Product Details