The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113182.2 | NP_001106653.1 | 8 - 9 | 2941 | 66 |
NM_001199455.1 | NP_001186384.1 | 7 - 8 | 2631 | 66 | |
NM_001199456.1 | NP_001186385.1 | 7 - 8 | 1344 | 66 | |
NM_001291986.1 | NP_001278915.1 | 9 - 10 | 3033 | 66 | |
NM_005104.3 | NP_005095.1 | 8 - 9 | 3028 | 66 | |
GenBank mRNA | AK056504.1 | - | 8 - 9 | 3081 | 66 |
AL832722.1 | - | 9 - 10 | 2968 | 66 | |
BC000477.2 | - | 7 - 8 | 2000 | 66 | |
BC063840.1 | - | 7 - 8 | 1496 | 66 | |
BX647233.1 | - | 7 - 8 | 1344 | 66 | |
BX648109.1 | - | 8 - 9 | 2942 | 66 | |
D42040.1 | - | 8 - 9 | 3028 | 66 | |
M80613.1 | - | 7 - 8 | 2363 | 66 | |
X62083.1 | - | 7 - 8 | 2363 | 66 |