The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001526.3 | NP_001517.2 | 1 - 2 | 533 | 68 |
XM_017010798.1 | XP_016866287.1 | 1 - 2 | 488 | 68 | |
GenBank mRNA | AF041245.1 | - | 1 - 2 | 577 | 68 |
AK314279.1 | - | 1 - 2 | 485 | 68 | |
KC812499.1 | - | 2 - 3 | 681 | 68 | |
KC812500.1 | - | 2 - 3 | 681 | 68 |