The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289034.1 | NP_001275963.1 | 4 - 5 | 923 | 79 |
NM_001289035.1 | NP_001275964.1 | 3 - 4 | 458 | 79 | |
NM_138343.3 | NP_612352.1 | 4 - 5 | 689 | 79 | |
NM_201521.2 | NP_958929.1 | 4 - 5 | 689 | 79 | |
NM_201522.2 | NP_958930.1 | 5 - 6 | 1105 | 79 | |
NM_201523.2 | NP_958931.1 | 4 - 5 | 979 | 79 | |
XM_011514989.1 | XP_011513291.1 | 4 - 5 | 1038 | 79 | |
XM_017011457.1 | XP_016866946.1 | 4 - 5 | 689 | 79 | |
GenBank mRNA | AK055293.1 | - | 5 - 6 | 1067 | 79 |
AK056393.1 | - | 4 - 5 | 649 | 79 | |
AK094102.1 | - | 4 - 5 | 945 | 79 | |
AK127018.1 | - | 7 - 8 | 1954 | 79 | |
AK127894.1 | - | 4 - 5 | 662 | 79 | |
AK297433.1 | - | 3 - 4 | 421 | 79 | |
BC012357.1 | - | 4 - 5 | 630 | 79 | |
BC080637.1 | - | 4 - 5 | 629 | 79 | |
BC103727.1 | - | 4 - 5 | 627 | 79 | |
BX640717.1 | - | 4 - 5 | 886 | 79 |