The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289034.1 | NP_001275963.1 | 6 - 7 | 1232 | 70 |
NM_001289035.1 | NP_001275964.1 | 5 - 6 | 767 | 70 | |
NM_201521.2 | NP_958929.1 | 6 - 7 | 998 | 70 | |
NM_201522.2 | NP_958930.1 | 7 - 8 | 1414 | 70 | |
NM_201523.2 | NP_958931.1 | 6 - 7 | 1288 | 70 | |
XM_011514989.1 | XP_011513291.1 | 6 - 7 | 1347 | 70 | |
XM_017011457.1 | XP_016866946.1 | 6 - 7 | 998 | 70 | |
GenBank mRNA | AK055293.1 | - | 7 - 8 | 1376 | 70 |
AK094102.1 | - | 6 - 7 | 1254 | 70 | |
AK127018.1 | - | 9 - 10 | 2263 | 70 | |
AK127894.1 | - | 6 - 7 | 971 | 70 | |
AK297433.1 | - | 5 - 6 | 730 | 70 | |
AL162078.1 | - | 1 - 2 | 89 | 70 | |
BC080637.1 | - | 6 - 7 | 938 | 70 | |
BC103727.1 | - | 6 - 7 | 936 | 70 | |
BX640717.1 | - | 6 - 7 | 1195 | 70 |