The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289034.1 | NP_001275963.1 | 9 - 10 | 1603 | 62 |
NM_001289035.1 | NP_001275964.1 | 8 - 9 | 1138 | 62 | |
NM_201521.2 | NP_958929.1 | 9 - 10 | 1369 | 62 | |
NM_201522.2 | NP_958930.1 | 10 - 11 | 1785 | 62 | |
NM_201523.2 | NP_958931.1 | 9 - 10 | 1659 | 62 | |
XM_011514989.1 | XP_011513291.1 | 9 - 10 | 1718 | 62 | |
XM_017011457.1 | XP_016866946.1 | 9 - 10 | 1369 | 62 | |
GenBank mRNA | AK055293.1 | - | 10 - 11 | 1747 | 62 |
AK056393.1 | - | 9 - 10 | 1329 | 62 | |
AK094102.1 | - | 9 - 10 | 1625 | 62 | |
AK127894.1 | - | 9 - 10 | 1342 | 62 | |
AK297433.1 | - | 8 - 9 | 1101 | 62 | |
AL162078.1 | - | 4 - 5 | 460 | 62 | |
BC080637.1 | - | 9 - 10 | 1309 | 62 | |
BC103727.1 | - | 9 - 10 | 1307 | 62 | |
BX640717.1 | - | 9 - 10 | 1566 | 62 |