Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr.6: 42963873 - 42979243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000287.3 NP_000278.3 10 - 11 2187 52
NM_001316313.1 NP_001303242.1 10 - 11 1923 52
XM_011514661.1 XP_011512963.1 9 - 10 2103 52
GenBank mRNA AB051076.1 - 10 - 11 2094 52
AB051077.1 - 10 - 11 1830 52
AB208906.1 - 10 - 11 2147 52
AK314237.1 - 10 - 11 2123 52
BC033487.1 - 10 - 11 2102 52
BC048331.1 - 10 - 11 2110 52
D83703.1 - 10 - 11 2164 52

View Full Product Details