The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000287.3 | NP_000278.3 | 16 - 17 | 2892 | 92 |
NM_001316313.1 | NP_001303242.1 | 16 - 17 | 2628 | 92 | |
NR_133009.1 | 14 - 15 | 2645 | 92 | ||
XM_011514661.1 | XP_011512963.1 | 15 - 16 | 2808 | 92 | |
GenBank mRNA | AB051076.1 | - | 16 - 17 | 2799 | 92 |
AB051077.1 | - | 16 - 17 | 2535 | 92 | |
AB051078.1 | - | 14 - 15 | 2552 | 92 | |
AB208906.1 | - | 12 - 13 | 3486 | 92 | |
AK314237.1 | - | 16 - 17 | 2828 | 92 | |
BC033487.1 | - | 16 - 17 | 2807 | 92 | |
BC048331.1 | - | 16 - 17 | 2815 | 92 | |
D83703.1 | - | 16 - 17 | 2869 | 92 |