The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018034.2 | NP_060504.1 | 10 - 11 | 1151 | 81 |
XM_005248318.3 | XP_005248375.1 | 10 - 11 | 1169 | 81 | |
XM_011514061.2 | XP_011512363.1 | 4 - 5 | 464 | 81 | |
XM_017009604.1 | XP_016865093.1 | 9 - 10 | 985 | 81 | |
XM_017009605.1 | XP_016865094.1 | 9 - 10 | 952 | 81 | |
GenBank mRNA | AK001095.1 | - | 10 - 11 | 1093 | 81 |
AK291179.1 | - | 10 - 11 | 1173 | 81 | |
AL512685.1 | - | 10 - 11 | 1152 | 81 | |
BC009648.1 | - | 10 - 11 | 1151 | 81 | |
BC025315.1 | - | 10 - 11 | 1129 | 81 | |
BX537655.1 | - | 9 - 10 | 1108 | 81 |