The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018034.2 | NP_060504.1 | 4 - 5 | 356 | 168 |
XM_005248318.3 | XP_005248375.1 | 4 - 5 | 374 | 168 | |
GenBank mRNA | AK001095.1 | - | 4 - 5 | 298 | 168 |
AK291179.1 | - | 4 - 5 | 378 | 168 | |
AK310901.1 | - | 4 - 5 | 381 | 168 | |
BC009648.1 | - | 4 - 5 | 356 | 168 | |
BC025315.1 | - | 4 - 5 | 334 | 168 | |
BX537655.1 | - | 3 - 4 | 313 | 168 |