The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032476.3 | NP_115865.1 | 2 - 3 | 360 | 90 |
GenBank mRNA | AB049942.1 | - | 2 - 3 | 306 | 90 |
AK096374.1 | - | 2 - 3 | 1327 | 90 | |
AK312084.1 | - | 2 - 3 | 305 | 90 | |
AY061855.1 | - | 2 - 3 | 360 | 90 | |
BC000547.2 | - | 2 - 3 | 292 | 90 | |
BC004976.1 | - | 2 - 3 | 294 | 90 | |
BC010076.2 | - | 2 - 3 | 294 | 90 | |
BC051302.1 | - | 2 - 3 | 201 | 90 | |
BG720377.1 | - | 2 - 3 | 310 | 90 |