The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006547.2 | NP_006538.2 | 3 - 4 | 558 | 97 |
GenBank mRNA | AF117108.1 | - | 3 - 4 | 558 | 97 |
BC065269.1 | - | 3 - 4 | 537 | 97 | |
U76705.1 | - | 3 - 4 | 542 | 97 | |
U97188.1 | - | 3 - 4 | 542 | 97 |