The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284301.2 | NP_001271230.1 | 10 - 11 | 2126 | 96 |
NM_001284302.2 | NP_001271231.1 | 10 - 11 | 2123 | 96 | |
NM_018685.4 | NP_061155.2 | 11 - 12 | 2237 | 96 | |
XM_006715746.1 | XP_006715809.1 | 11 - 12 | 2234 | 96 | |
XM_006715747.3 | XP_006715810.1 | 10 - 11 | 2123 | 96 | |
XM_017012354.1 | XP_016867843.1 | 11 - 12 | 2222 | 96 | |
XM_017012355.1 | XP_016867844.1 | 11 - 12 | 2222 | 96 | |
XM_017012356.1 | XP_016867845.1 | 10 - 11 | 2111 | 96 | |
GenBank mRNA | AF273437.1 | - | 11 - 12 | 2172 | 96 |
AK001468.1 | - | 1 - 2 | 156 | 96 | |
AK001472.1 | - | 6 - 7 | 904 | 96 | |
AK023208.1 | - | 2 - 3 | 278 | 96 | |
AK291254.1 | - | 10 - 11 | 2055 | 96 | |
AK299803.1 | - | 11 - 12 | 1939 | 96 | |
BC070066.1 | - | 10 - 11 | 2108 | 96 | |
CR936650.1 | - | 8 - 9 | 1327 | 96 |