Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2
Entrez Gene ID
140683
Gene Name
BPI fold containing family A member 2
Gene Aliases
C20orf70, PSP, SPLUNC2, bA49G10.1
Chromosome Location
Chr.20: 33161848 - 33181417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319164.1 NP_001306093.1 2 - 3 228 107
NM_080574.3 NP_542141.1 2 - 3 310 107
GenBank mRNA AF432917.1 - 1 - 2 162 107
AY359055.1 - 2 - 3 228 107
BC065726.1 - 2 - 3 237 107

View Full Product Details