The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014819.4 | NP_055634.3 | 9 - 10 | 2224 | 78 |
XM_017010099.1 | XP_016865588.1 | 10 - 11 | 2300 | 78 | |
GenBank mRNA | AB007898.1 | - | 9 - 10 | 2121 | 78 |
AK291759.1 | - | 9 - 10 | 2158 | 78 | |
BC030826.1 | - | 9 - 10 | 2120 | 78 | |
CR749579.1 | - | 5 - 6 | 581 | 78 |