Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC93A
Entrez Gene ID
54346
Gene Name
unc-93 homolog A (C. elegans)
Gene Aliases
Unc-93A, dJ366N23.1, dJ366N23.2
Chromosome Location
Chr.6: 167269032 - 167316014 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143947.1 NP_001137419.1 2 - 3 446 67
NM_018974.3 NP_061847.2 2 - 3 446 67
XM_011535905.2 XP_011534207.1 3 - 4 852 67
XM_011535906.2 XP_011534208.1 4 - 5 1001 67
XM_011535907.2 XP_011534209.1 3 - 4 1076 67
XM_011535908.2 XP_011534210.1 2 - 3 446 67
XM_017010958.1 XP_016866447.1 3 - 4 417 67
GenBank mRNA AJ422141.1 - 2 - 3 374 67
AJ508812.1 - 2 - 3 348 67
AK091987.1 - 2 - 3 446 67
BC098248.1 - 2 - 3 297 67
BC099718.1 - 2 - 3 297 67
BC105635.1 - 2 - 3 297 67

View Full Product Details