The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015978.2 | NP_057062.1 | 4 - 5 | 400 | 74 |
GenBank mRNA | AF116826.1 | - | 4 - 5 | 386 | 74 |
AK307984.1 | - | 4 - 5 | 381 | 74 | |
AY303691.1 | - | 4 - 5 | 365 | 74 | |
BC113539.1 | - | 4 - 5 | 374 | 74 | |
BC117262.1 | - | 4 - 5 | 374 | 74 | |
BX502807.1 | - | 4 - 5 | 401 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112808.2 | NP_001106279.2 | 6 - 7 | 705 | 74 |
NM_001199327.1 | NP_001186256.2 | 6 - 7 | 705 | 74 | |
GenBank mRNA | AB208941.1 | - | 6 - 7 | 654 | 74 |
BC032865.1 | - | 6 - 7 | 678 | 74 | |
BX640903.1 | - | 6 - 7 | 690 | 74 |