The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282143.1 | NP_001269072.1 | 5 - 6 | 1086 | 92 |
NM_001282144.1 | NP_001269073.1 | 5 - 6 | 1494 | 92 | |
NM_001282358.1 | NP_001269287.1 | 5 - 6 | 1018 | 92 | |
NM_024618.3 | NP_078894.2 | 5 - 6 | 1113 | 92 | |
XM_005271669.1 | XP_005271726.1 | 5 - 6 | 1337 | 92 | |
XM_005271670.1 | XP_005271727.1 | 4 - 5 | 990 | 92 | |
XM_005271671.1 | XP_005271728.1 | 4 - 5 | 1164 | 92 | |
XM_005271672.1 | XP_005271729.1 | 4 - 5 | 987 | 92 | |
XM_006718904.1 | XP_006718967.1 | 5 - 6 | 1371 | 92 | |
XM_011542980.1 | XP_011541282.1 | 5 - 6 | 1001 | 92 | |
XM_017018294.1 | XP_016873783.1 | 5 - 6 | 1371 | 92 | |
GenBank mRNA | AB094095.1 | - | 5 - 6 | 1097 | 92 |
AK056454.1 | - | 4 - 5 | 811 | 92 | |
AK095247.1 | - | 3 - 4 | 298 | 92 | |
AK291716.1 | - | 5 - 6 | 1080 | 92 | |
AK303002.1 | - | 4 - 5 | 987 | 92 | |
AY245437.1 | - | 4 - 5 | 856 | 92 | |
BC013199.1 | - | 1 - 2 | 217 | 92 | |
BC110890.1 | - | 5 - 6 | 1459 | 92 | |
BX647705.1 | - | 5 - 6 | 1071 | 92 | |
EF452237.1 | - | 4 - 5 | 856 | 92 |