The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005441.2 | NP_005432.1 | 9 - 10 | 965 | 106 |
XM_011529755.2 | XP_011528057.1 | 8 - 9 | 904 | 106 | |
XM_017028477.1 | XP_016883966.1 | 11 - 12 | 1858 | 106 | |
XM_017028478.1 | XP_016883967.1 | 11 - 12 | 2234 | 106 | |
GenBank mRNA | AK313152.1 | - | 9 - 10 | 960 | 106 |
BC001143.1 | - | 9 - 10 | 941 | 106 | |
BC021218.2 | - | 8 - 9 | 897 | 106 | |
U20980.1 | - | 8 - 9 | 889 | 106 |