The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032587.3 | NP_115976.2 | 2 - 3 | 908 | 103 |
GenBank mRNA | AB209844.1 | - | 2 - 3 | 728 | 103 |
AF356193.1 | - | 2 - 3 | 1032 | 103 | |
AK304349.1 | - | 2 - 3 | 840 | 103 | |
BC093825.1 | - | 2 - 3 | 855 | 103 |