Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM4
Entrez Gene ID
9202
Gene Name
zinc finger MYM-type containing 4
Gene Aliases
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location
Chr.1: 35268694 - 35421944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005271331.2 XP_005271388.1 2 - 3 166 109
XM_011542425.2 XP_011540727.1 3 - 4 206 109
GenBank mRNA AB007885.1 - 1 - 2 41 109
BC127114.1 - 2 - 3 110 109

View Full Product Details