The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030815.2 | NP_110442.1 | 4 - 5 | 432 | 146 |
GenBank mRNA | AY286301.1 | - | 4 - 5 | 444 | 146 |
BC001856.2 | - | 4 - 5 | 303 | 146 | |
BC009334.2 | - | 4 - 5 | 357 | 146 |