The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197330.1 | NP_001184259.1 | 8 - 9 | 779 | 77 |
NM_001197331.1 | NP_001184260.1 | 9 - 10 | 857 | 77 | |
NM_002692.3 | NP_002683.2 | 9 - 10 | 857 | 77 | |
XM_011536842.2 | XP_011535144.1 | 8 - 9 | 816 | 77 | |
XM_011536843.2 | XP_011535145.1 | 9 - 10 | 694 | 77 | |
XM_011536844.1 | XP_011535146.1 | 9 - 10 | 691 | 77 | |
XM_017021374.1 | XP_016876863.1 | 9 - 10 | 691 | 77 | |
XM_017021375.1 | XP_016876864.1 | 9 - 10 | 694 | 77 | |
GenBank mRNA | AF025840.1 | - | 9 - 10 | 810 | 77 |
AF036899.1 | - | 9 - 10 | 694 | 77 | |
AK293163.1 | - | 8 - 9 | 611 | 77 | |
AK314963.1 | - | 9 - 10 | 692 | 77 | |
BC112962.1 | - | 9 - 10 | 687 | 77 | |
BC126218.1 | - | 9 - 10 | 780 | 77 | |
BC126220.1 | - | 9 - 10 | 780 | 77 | |
BX248064.1 | - | 8 - 9 | 617 | 77 |