The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178030.1 | NP_001171501.1 | 10 - 11 | 1798 | 64 |
NM_001178031.1 | NP_001171502.1 | 10 - 11 | 1580 | 64 | |
NM_004630.3 | NP_004621.2 | 10 - 11 | 1801 | 64 | |
NM_201995.2 | NP_973724.1 | 10 - 11 | 1801 | 64 | |
NM_201998.2 | NP_973727.1 | 10 - 11 | 1801 | 64 | |
NR_033649.1 | 9 - 10 | 1725 | 64 | ||
XM_011545244.1 | XP_011543546.1 | 10 - 11 | 1473 | 64 | |
XM_017018244.1 | XP_016873733.1 | 10 - 11 | 1473 | 64 | |
XM_017018245.1 | XP_016873734.1 | 10 - 11 | 1554 | 64 | |
XM_017018249.1 | XP_016873738.1 | 10 - 11 | 3966 | 64 | |
XM_017018250.1 | XP_016873739.1 | 9 - 10 | 1650 | 64 | |
XM_017018251.1 | XP_016873740.1 | 9 - 10 | 1505 | 64 | |
GenBank mRNA | AK293753.1 | - | 10 - 11 | 1580 | 64 |
AK296237.1 | - | 10 - 11 | 1472 | 64 | |
AK299705.1 | - | 8 - 9 | 1211 | 64 | |
AK301803.1 | - | 9 - 10 | 1650 | 64 | |
BC000773.1 | - | 1 - 2 | 229 | 64 | |
BC008080.2 | - | 10 - 11 | 1497 | 64 | |
BC008724.2 | - | 10 - 11 | 1497 | 64 | |
BC020217.1 | - | 10 - 11 | 1700 | 64 | |
BC034451.1 | - | 10 - 11 | 1726 | 64 | |
BC038446.1 | - | 10 - 11 | 1821 | 64 | |
D26120.1 | - | 10 - 11 | 1731 | 64 | |
L49380.1 | - | 10 - 11 | 1687 | 64 | |
Y08765.1 | - | 10 - 11 | 1391 | 64 | |
Y08766.1 | - | 10 - 11 | 1643 | 64 |