Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPX
Entrez Gene ID
80763
Gene Name
spexin hormone
Gene Aliases
C12orf39
Chromosome Location
Chr.12: 21526257 - 21532947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030572.3 NP_085049.1 3 - 4 362 64
GenBank mRNA AK027273.1 - 3 - 4 200 64
AK075342.1 - 3 - 4 316 64
BC004336.1 - 3 - 4 297 64

View Full Product Details