Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 19 - 20 3256 56
NM_002845.3 NP_002836.3 17 - 18 3217 56
XM_011525708.2 XP_011524010.1 20 - 21 3326 56
XM_011525710.2 XP_011524012.1 20 - 21 3326 56
XM_011525712.2 XP_011524014.1 19 - 20 3251 56
XM_011525713.2 XP_011524015.1 20 - 21 2926 56
XM_011525715.2 XP_011524017.1 16 - 17 2291 56
XM_011525716.2 XP_011524018.1 13 - 14 2171 56
XM_011525717.2 XP_011524019.1 14 - 15 2000 56
XM_011525722.2 XP_011524024.1 15 - 16 2216 56
XM_017025894.1 XP_016881383.1 20 - 21 2870 56
XM_017025895.1 XP_016881384.1 19 - 20 2858 56
XM_017025896.1 XP_016881385.1 18 - 19 2831 56
XM_017025897.1 XP_016881386.1 20 - 21 2870 56
XM_017025898.1 XP_016881387.1 19 - 20 2858 56
XM_017025899.1 XP_016881388.1 19 - 20 2795 56
XM_017025900.1 XP_016881389.1 18 - 19 2831 56
XM_017025901.1 XP_016881390.1 17 - 18 2756 56
XM_017025902.1 XP_016881391.1 19 - 20 2795 56
XM_017025903.1 XP_016881392.1 18 - 19 2783 56
XM_017025904.1 XP_016881393.1 17 - 18 2756 56
XM_017025905.1 XP_016881394.1 20 - 21 2926 56
XM_017025906.1 XP_016881395.1 16 - 17 2275 56
XM_017025907.1 XP_016881396.1 15 - 16 2216 56
XM_017025908.1 XP_016881397.1 14 - 15 2252 56
XM_017025909.1 XP_016881398.1 14 - 15 2204 56
GenBank mRNA AK294492.1 - 13 - 14 2098 56
BC051651.1 - 17 - 18 2953 56
BC151842.1 - 19 - 20 3802 56
X58288.1 - 17 - 18 2714 56

View Full Product Details